Merck
  • 137 differential gene expression of in vitro-matured bovine oocytes with or without a polar body.
137 differential gene expression of in vitro-matured bovine oocytes with or without a polar body.
Reproduction, Fertility, and Development (2011)
M. M. Pereira et al.