Merck
  • Partial molar volumes of transfer of some disaccharides from water to aqueous guanidine hydrochloride solutions at 298.15 K.
Partial molar volumes of transfer of some disaccharides from water to aqueous guanidine hydrochloride solutions at 298.15 K.
Indian J. Chem. A (2002)
Banipal TS, et al.