- Multiplex PCR for rapid detection of genes encoding acquired metallo-beta-lactamases.
Multiplex PCR for rapid detection of genes encoding acquired metallo-beta-lactamases.
The Journal of antimicrobial chemotherapy (2006-12-23)
Matthew J Ellington, James Kistler, David M Livermore, Neil Woodford
PMID17185300
MATERIALIEN