Merck
  • cAMP regulates expression of the cyclic nucleotide transporter MRP4 (ABCC4) through the EPAC pathway.
cAMP regulates expression of the cyclic nucleotide transporter MRP4 (ABCC4) through the EPAC pathway.
Pharmacogenetics and Genomics (2014)
Broderdorf S, Zang S, Schaletzki Y, et al.