Merck
  • Frustrated Lewis pair catalyzed hydrosilylation and hydrosilane mediated hydrogenation of fulvenes.
Frustrated Lewis pair catalyzed hydrosilylation and hydrosilane mediated hydrogenation of fulvenes.
Organic & biomolecular chemistry (2014-10-09)
Sergej Tamke, Constantin-G Daniliuc, Jan Paradies
ZUSAMMENFASSUNG
The frustrated Lewis pair (FLP) mediated hydrosilylation of pentafulvenes is described yielding allyl silanes with high regioselectivity in excellent yields. While phenyl substituted allyl silanes undergo B(C6F5)3-mediated rearrangement to vinyl silanes, dimethyl derivatives experience FLP-catalyzed hydrogenation followed by an unprecedented protodesilylation. This observation allowed the metal-free hydrogenation of 6,6-dimethylfulvene to iso-propyl cyclopentene according to a FLP-catalyzed triple domino reaction consisting of hydrosilylation, hydrogenation and protodesilylation. The mechanisms were investigated by deuteration experiments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6,6-Dimethylfulven, ≥95%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.