Merck
  • A novel polymorphic allele of human arylacetamide deacetylase leads to decreased enzyme activity.
A novel polymorphic allele of human arylacetamide deacetylase leads to decreased enzyme activity.
Drug metabolism and disposition: the biological fate of chemicals (2012-03-15)
Mai Shimizu, Tatsuki Fukami, Yuki Kobayashi, Masataka Takamiya, Yasuhiro Aoki, Miki Nakajima, Tsuyoshi Yokoi
ZUSAMMENFASSUNG
Human arylacetamide deacetylase (AADAC) is responsible for the hydrolysis of clinically used drugs such as flutamide, phenacetin, and rifamycins. Our recent studies suggested that human AADAC is a relevant enzyme pharmacologically and toxicologically. To date, the genetic polymorphisms that affect enzyme activity in AADAC have been unknown. In this study, we found single-nucleotide polymorphisms in the human AADAC gene in a liver sample that showed remarkably low flutamide hydrolase activity. Among them, g.13651G > A (V281I) and g.14008T > C (X400Q) were nonsynonymous. The latter would be predicted to cause a C-terminal one-amino acid (glutamine) extension. The AADAC*2 allele (g.13651G > A) was found in all populations investigated in this study (European American, African American, Korean, and Japanese), at allelic frequencies of 52.6 to 63.5%, whereas the AADAC*3 allele (g.13651G > A/g.14008T > C) was found in European American (1.3%) and African American (2.0%) samples. COS7 cells expressing AADAC.1 (wild-type) exhibited flutamide, phenacetin, and rifampicin hydrolase activities with intrinsic clearance (CLint) values of 1.31 ± 0.06, 1.00 ± 0.02, and 0.39 ± 0.02 μl x min(-1) x unit(-1), respectively. AADAC.2, which is a protein produced from the AADAC*2 allele, showed moderately lower or similar CLint values, compared with AADAC.1, but AADAC.3 showed substantially lower CLint values (flutamide hydrolase, 0.21 ± 0.02 μl x min(-1) x unit(-1); phenacetin hydrolase, 0.12 ± 0.00 μl x min(-1) x unit(-1); rifampicin hydrolase, 0.03 ± 0.01 μl x min(-1) x unit(-1), respectively). Microsomes from a liver sample genotyped as AADAC*3/AADAC*3 showed decreased enzyme activities, compared with those genotyped as AADAC*1/AADAC*1, AADAC*1/AADAC*2, and AADAC*2/AADAC*2. In conclusion, we found an AADAC allele that yielded decreased enzyme activity. This study should provide useful information on interindividual variations in AADAC enzyme activity.