Merck
  • Synthesis of rhenium-based M 2 LL'-type supramolecular coordination complexes from flexible ligands.
Synthesis of rhenium-based M 2 LL'-type supramolecular coordination complexes from flexible ligands.
Journal of Organometallic Chemistry (2013)
Shankar B, et al.