Merck
  • Occurrence and spatial distribution of EDCs and related compounds in waters and sediments of Iberian rivers.
Occurrence and spatial distribution of EDCs and related compounds in waters and sediments of Iberian rivers.
The Science of the total environment (2014-07-16)
Marina Gorga, Sara Insa, Mira Petrovic, Damià Barceló
ZUSAMMENFASSUNG
The environmental presence of chemicals capable of affecting the endocrine system has become a matter of scientific and public concern after certain endocrine disruptor compounds (EDCs) have been detected in the aquatic environment. In this work, 31 different EDCs and related compounds (suspect EDCs) belonging to different contaminant classes were studied: 10 estrogens, natural and synthetic, 8 alkylphenolic compounds, bisphenol A, triclosan and triclorocaraban, 4 parabens, 2 benzotriazoles, 3 organophosphorous flame retardants and the chemical marker caffeine, in river water and sediment of four Iberian rivers (Ebro, Llobregat, Júcar and Guadalquivir). An extensive sampling has been undertaken in two monitoring campaigns (2010 and 2011). A total of 77 samples of water and 75 sediments were collected. For this propose two different multiresidue analytical methods were applied, using the automated online EQuan/TurboFlow™ liquid chromatography coupled to mass spectrometry detection in tandem. In terms of concentrations the compounds found at the highest average concentrations were: nonylphenol monocarboxylate (NP1EC), tolyltriazole (TT), tris(chloroisopropyl)phosphate (TCPP) found at average concentrations above 100 ng/L, followed by 1H-benzotriazole and tris(butoxyethyl)phosphate (TBEP) found at average concentration higher than 50 ng/L. Natural and synthetic hormones were found at low levels not exceeding 16 ng/L and 7 ng/g for water and sediment, respectively, however they contributed to more than 80% of the total estrogenicity of the samples (expressed as the equivalents of estradiol EEQ, ng/L). Regarding the spatial distribution of these contaminants, the Llobregat river was found to be the most contaminated river basin, having sites near the mouth of the river the ones with the highest contaminant load. In the Ebro river basin several hot spots were identified and Júcar showed to be the least contaminated. Overall, the study confirmed the presence of complex mixtures of unregulated contaminants, thus raising concern about their potential interactive effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ameisensäure, reagent grade, ≥95%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥96%
Supelco
Ammoniumformat, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammoniumformat, reagent grade, 97%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Ammoniumformat, ≥99.995% trace metals basis
Sigma-Aldrich
Ameisensäure, ACS reagent, ≥88%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammoniumformiat -Lösung, BioUltra, 10 M in H2O
Supelco
4,4′-Isopropylidendiphenol, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammoniumformat, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
4,4′-Isopropylidendiphenol, 97%
Sigma-Aldrich
Nonylphenol, technical grade, mixture of ring and chain isomers
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
4-tert-Octylphenol, 97%
Supelco
4,4′-Isopropylidendiphenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ethylsalicylat, 99%
Supelco
Nonylphenol, PESTANAL®, analytical standard, technical mixture