Merck
  • Lenalidomide for second-line treatment of advanced hepatocellular cancer: a Brown University oncology group phase II study.
Lenalidomide for second-line treatment of advanced hepatocellular cancer: a Brown University oncology group phase II study.
American journal of clinical oncology (2013-05-08)
Howard Safran, Kevin P Charpentier, Andreas Kaubisch, Kalyan Mantripragada, Gregory Dubel, Kimberly Perez, Katherine Faricy-Anderson, Thomas Miner, Yoko Eng, Joel Victor, Angela Plette, Joseph Espat, Pamela Bakalarski, Patti Wingate, David Berz, Denise Luppe, Diane Martel, Kayla Rosati, Santiago Aparo
ZUSAMMENFASSUNG
To assess the activity and toxicity of lenalidomide for patients with advanced hepatocellular cancer (HCC) previously treated with sorafenib. Patients with advanced HCC who progressed on or were intolerant to sorafenib were eligible. Patients received lenalidomide 25 mg orally for 1 to 21 days in a 28-day cycle until disease progression or unacceptable toxicities. Forty patients were enrolled and were classified according to the Child-Pugh score: 19 were Child-Pugh A, 16 patients were Child-Pugh B, and 5 were Child-Pugh C. Seventeen patients had extrahepatic disease. Grade 4 neutropenia occurred in 1 of 40 patients (2.5%). Grade 3 fatigue (n=3) and rash (n=4) were the most common nonhematologic toxicities attributable to lenalidomide. Six of 40 patients (15%) had a partial response. Two patients (5%) have not progressed at 36 and 32 months. The median progression-free survival was 3.6 months and the median overall survival was 7.6 months. Lenalidomide can be administered to patients with advanced HCC and hepatic dysfunction. Promising, and in a small percentage of patients, durable activity has been demonstrated. Investigations are needed to explore the mechanism of action of lenalidomide in HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nicotinamid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamid, ≥99.5% (HPLC)
Supelco
Niacinamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Nicotinamid, ≥98% (HPLC), powder
USP
Niacinamid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamid, meets USP testing specifications
Sigma-Aldrich
(±)-Thalidomid, ≥98%, powder
Supelco
Nicotinamid (Vitamin B3) -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamid, ≥98.5% (HPLC)
Nicotinamid, European Pharmacopoeia (EP) Reference Standard