Merck
  • Klotho gene deficiency causes salt-sensitive hypertension via monocyte chemotactic protein-1/CC chemokine receptor 2-mediated inflammation.
Klotho gene deficiency causes salt-sensitive hypertension via monocyte chemotactic protein-1/CC chemokine receptor 2-mediated inflammation.
Journal of the American Society of Nephrology : JASN (2014-06-07)
Xiaoli Zhou, Kai Chen, Han Lei, Zhongjie Sun
ZUSAMMENFASSUNG
Klotho (KL) is a newly discovered aging suppressor gene. In mice, the KL gene extends the lifespan when overexpressed and shortens the lifespan when disrupted. This study investigated if KL deficiency affects BP and salt sensitivity using KL mutant heterozygous (+/-) mice and wild-type (WT) mice (9 weeks of age, 16 mice per group). Notably, systolic BP in KL(+/-) mice began to increase at the age of 15 weeks, reached a peak level at the age of 17 weeks, and remained elevated thereafter, whereas systolic BP remained consistent in WT mice. High salt (HS) intake further increased BP in KL(+/-) mice but did not affect BP in WT mice. Blockade of CC chemokine receptor 2 (CCR2), involved in monocyte chemotaxis, by a specific CCR2 antagonist (INCB3284) abolished the HS-induced increase in BP in KL(+/-) mice. Furthermore, HS loading substantially increased the expression of monocyte chemotactic protein-1 and the infiltration of macrophages and T cells in kidneys in KL(+/-) mice, and treatment with INCB3284 abolished these effects. Treatment of KL(+/-) mice with INCB3284 also attenuated the increased renal expressions of serum glucocorticoid-regulated kinase 1, thiazide-sensitive NaCl cotransporter, and ATP synthase β along with the renal structural damage and functional impairment induced by HS loading. In conclusion, KL deficiency caused salt-sensitive hypertension and renal damage by CCR2-mediated inflammation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycin -hydrochlorid, ≥99% (HPLC)
SAFC
Glycin
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, 99%, FCC
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycin 1 M -Lösung
Supelco
Glycin, analytical standard, for nitrogen determination according to Kjeldahl method
Glycin, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycin -Lösung, 100 mM amino acid in 0.1 M HCl, analytical standard
Supelco
Glycin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycin, tested according to Ph. Eur.