Merck
  • Glucose-dependent insulinotropic polypeptide augments glucagon responses to hypoglycemia in type 1 diabetes.
Glucose-dependent insulinotropic polypeptide augments glucagon responses to hypoglycemia in type 1 diabetes.
Diabetes (2014-07-24)
Mikkel Christensen, Salvatore Calanna, Alexander H Sparre-Ulrich, Peter L Kristensen, Mette M Rosenkilde, Jens Faber, Francesco Purrello, Gerrit van Hall, Jens J Holst, Tina Vilsbøll, Filip K Knop
ZUSAMMENFASSUNG
Glucose-dependent insulinotropic polypeptide (GIP) is glucagonotropic, and glucagon-like peptide-1 (GLP-1) is glucagonostatic. We studied the effects of GIP and GLP-1 on glucagon responses to insulin-induced hypoglycemia in patients with type 1 diabetes mellitus (T1DM). Ten male subjects with T1DM (C-peptide negative, age [mean ± SEM] 26 ± 1 years, BMI 24 ± 0.5 kg/m(2), HbA1c 7.3 ± 0.2%) were studied in a randomized, double-blinded, crossover study, with 2-h intravenous administration of saline, GIP, or GLP-1. The first hour, plasma glucose was lowered by insulin infusion, and the second hour constituted a "recovery phase." During the recovery phase, GIP infusions elicited larger glucagon responses (164 ± 50 [GIP] vs. 23 ± 25 [GLP-1] vs. 17 ± 46 [saline] min ⋅ pmol/L, P < 0.03) and endogenous glucose production was higher with GIP and lower with GLP-1 compared with saline (P < 0.02). On the GIP days, significantly less exogenous glucose was needed to keep plasma glucose above 2 mmol/L (155 ± 36 [GIP] vs. 232 ± 40 [GLP-1] vs. 212 ± 56 [saline] mg ⋅ kg(-1), P < 0.05). Levels of insulin, cortisol, growth hormone, and noradrenaline, as well as hypoglycemic symptoms and cognitive function, were similar on all days. Our results suggest that during hypoglycemia in patients with T1DM, exogenous GIP increases glucagon responses during the recovery phase after hypoglycemia and reduces the need for glucose administration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, analytical standard
Sigma-Aldrich
Glycerin, tested according to Ph. Eur., anhydrous
Glucagon human, European Pharmacopoeia (EP) Reference Standard