Merck
  • Dietary pectin-derived acidic oligosaccharides improve the pulmonary bacterial clearance of Pseudomonas aeruginosa lung infection in mice by modulating intestinal microbiota and immunity.
Dietary pectin-derived acidic oligosaccharides improve the pulmonary bacterial clearance of Pseudomonas aeruginosa lung infection in mice by modulating intestinal microbiota and immunity.
The Journal of infectious diseases (2014-08-21)
Henry Bernard, Jean-Luc Desseyn, Nana Bartke, Lennart Kleinjans, Bernd Stahl, Clara Belzer, Jan Knol, Frédéric Gottrand, Marie-Odile Husson
ZUSAMMENFASSUNG
A predominantly T-helper type 2 (Th2) immune response is critical in the prognosis of pulmonary Pseudomonas aeruginosa infection. But the mucosal and systemic immune responses can be influenced by the intestinal microbiota. We assessed the effect of microbiota compositional changes induced by a diet enriched in 5% acidic oligosaccharides derived from pectin (pAOS) on the immune response and outcome of chronic pulmonary P. aeruginosa infection in mice. pAOS promoted Th1 polarization by increasing interferon γ release, upregulating t-bet gene expression, decreasing interleukin 4 secretion, and downregulating gata3 gene expression. pAOS also sustained the release of keratinocyte chemoattractant, recruited polynuclear leukocytes and macrophages, stimulated M1 macrophage activation and interleukin 10 release, and decreased tumor necrosis factor α release in the lung. These effects led to increased bacterial clearance after the first and second P. aeruginosa infections. pAOS modified the intestinal microbiota by stimulating the growth of species involved in immunity development, such as Bifidobacterium species, Sutturella wadsworthia, and Clostridium cluster XIVa organisms, and at the same time increased the production of butyrate and propionate. These results suggest that pAOS may have beneficial effects by limiting the number and severity of pulmonary exacerbations in patients chronically infected with P. aeruginosa, such as individuals with cystic fibrosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Buttersäure, ≥99%
Sigma-Aldrich
Propionsäure, ≥99.5%
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Pektin aus Citrusfrüchten (Schalen), Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
Valeriansäure, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 1.0 M in diethyl ether
Sigma-Aldrich
Isovaleriansäure, 99%
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Propionsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isobuttersäure, 99%
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Supelco
Propionsäure, analytical standard
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chlorwasserstoff – Methanol -Lösung, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Essigsäure, analytical standard