Merck
  • BDNF and Huntingtin protein modifications by manganese: implications for striatal medium spiny neuron pathology in manganese neurotoxicity.
BDNF and Huntingtin protein modifications by manganese: implications for striatal medium spiny neuron pathology in manganese neurotoxicity.
Journal of neurochemistry (2014-08-08)
Kirstie H Stansfield, Terry Jo Bichell, Aaron B Bowman, Tomás R Guilarte
ZUSAMMENFASSUNG
High levels of manganese (Mn) exposure decrease striatal medium spiny neuron (MSN) dendritic length and spine density, but the mechanism(s) are not known. The Huntingtin (HTT) gene has been functionally linked to cortical brain-derived neurotrophic factor (BDNF) support of striatal MSNs via phosphorylation at serine 421. In Huntington's disease, pathogenic CAG repeat expansions of HTT decrease synthesis and disrupt transport of cortical-striatal BDNF, which may contribute to disease, and Mn is a putative environmental modifier of Huntington's disease pathology. Thus, we tested the hypothesis that changes in MSN dendritic morphology Mn due to exposure are associated with decreased BDNF levels and alterations in Htt protein. We report that BDNF levels are decreased in the striatum of Mn-exposed non-human primates and in the cerebral cortex and striatum of mice exposed to Mn. Furthermore, proBDNF and mature BDNF concentrations in primary cortical and hippocampal neuron cultures were decreased by exposure to Mn confirming the in vivo findings. Mn exposure decreased serine 421 phosphorylation of Htt in cortical and hippocampal neurons and increased total Htt levels. These data strongly support the hypothesis that Mn-exposure-related MSN pathology is associated with decreased BDNF trophic support via alterations in Htt.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Anti-Huntingtinprotein-Antikörper, Klon mEM48, culture supernatant, clone mEM48, Chemicon®
Sigma-Aldrich
Anti-MAP2-Antikörper, Klon AP20, clone AP20, Chemicon®, from mouse
Sigma-Aldrich
Anti-Mikrotubuli-assoziiertes Protein 2 (MAP2)-Antikörper, Chemicon®, from rabbit
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Mangan, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Sigma-Aldrich
Anti-Brain Derived Neurotrophic Factor Antibody, pro, Chemicon®, from rabbit
Mangan, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mangan, rod, 50mm, diameter 4.0mm, cast, 99.5%
Mangan, foil, not light tested, 50x50mm, thickness 0.006mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.004mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.007mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.0025mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.008mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.02mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.004mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.006mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.007mm, permanent polyester support, 98.7%
Mangan, foil, not light tested, 25x25mm, thickness 0.009mm, permanent polyester support, 98.7%