Merck
  • Combining immunodetection with histochemical techniques: the effect of heat-induced antigen retrieval on picro-Sirius red staining.
Combining immunodetection with histochemical techniques: the effect of heat-induced antigen retrieval on picro-Sirius red staining.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (2014-09-14)
Victoria L Gadd
ZUSAMMENFASSUNG
Picro-Sirius red is a routine diagnostic stain intended for the histological visualization of collagen fibers (fibrosis) in tissue. Multi-label immunohistochemistry is a powerful tool used by researchers to visualize different cell types and their location within a tissue specimen, and to observe co-localization of antigens. Combining the specificity of immunodetection with the simplicity of Sirius red staining will allow researchers to visualize multi-antigen detection in relation to fibrosis, a common histological feature of injury in many chronic diseases. Pre-treatment of formalin-fixed, paraffin-embedded tissue (FFPE) specimens with antigen retrieval is essential for the work-up of most commercially available antibodies. The most common form of antigen retrieval involves boiling tissue specimens in buffer to break the cross-linkages caused by formalin fixation. However, this method causes tissue modification and collagen fiber shrinkage leading to suboptimal results when counterstaining for Sirius red. Reduced heat and enzymatic digestion are antigen retrieval methods compatible with Sirius red counterstaining. This paper will discuss the difficulties faced when combining these two staining methods, and provide a detailed method for the simultaneous detection of antigen and Sirius red in FFPE tissues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.