Merck
  • A sensitive monoclonal antibody-based enzyme-linked immunosorbent assay for the detection of bifenthrin in a chemical soil barrier.
A sensitive monoclonal antibody-based enzyme-linked immunosorbent assay for the detection of bifenthrin in a chemical soil barrier.
The Science of the total environment (2014-09-28)
Xiude Hua, Xiaofeng Liu, Wei Yin, Yazhong Xia, Qiujun Zhou, Yiwen Lu, Wei Li, Haiyan Shi, Fengquan Liu, Minghua Wang
ZUSAMMENFASSUNG
A sensitive monoclonal antibody-based enzyme monoclonal antibody-based enzyme-linked immunosorbent assay (ELISA) was developed to detect bifenthrin in a chemical soil barrier for termite control. LBc ((2-methyl[1,1-biphenyl]-3-methoxy) carbonyl propionic acid) was conjugated to bovine serum albumin (BSA) for producing monoclonal antibody. After optimization, the IC50 and limit of detection (LOD, IC10) were 0.05 mg L(-1) and 0.004 mg L(-1), respectively. The sensitivity was improved 40-fold compared to polyclonal antibody-based ELISA reported earlier. No cross-reactivity was measured for the other analogues such as cyhalothrin, cyhalothric acid, cypermethrin, deltamethrin, fenpropathrin, fenvalerate, ethofenprox and tetramethrin except 2-methyl-3-biphenylmethyanol. Spiked recoveries were between 83.5% and 104.7% for the detection of bifenthrin in loess, red soil and black soil. All the relative standard deviations (RSDs) were less than or equal to 15.0%. Moreover, the ELISA for authentic samples showed reliability and high correlation with gas chromatography. The developed ELISA is an alternative tool for simple, sensitive and accurate monitoring of bifenthrin in chemical soil barriers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Bifenthrin, PESTANAL®, analytical standard, mixture of isomers