Merck
  • Human biomonitoring after chemical incidents and during short-term maintenance work as a tool for exposure analysis and assessment.
Human biomonitoring after chemical incidents and during short-term maintenance work as a tool for exposure analysis and assessment.
Toxicology letters (2014-10-08)
M Bader, T Van Weyenbergh, E Verwerft, J Van Pul, S Lang, C Oberlinner
ZUSAMMENFASSUNG
Human biomonitoring (HBM) is frequently used for the analysis and assessment of exposure to chemicals under routine working conditions. In recent years, HBM has also been applied to monitor the exposure of the general population, and of emergency responders in the aftermath of chemical incidents. Two examples of targeted HBM programs in the chemical industry are described and discussed in this paper: (1) analysis and assessment of the exposure of firefighters and chemical workers after the spill of p-chloroaniline from a burning chemical barrel, and (2) biomonitoring of maintenance workers potentially exposed to benzene during regular turnarounds. The results of these investigations underline that human biomonitoring contributes substantially to comprehensive exposure analyses, human health risk assessments and communication. In addition, regular HBM surveillance and feedback can assist in the continuous improvement of workplace safety measures and exposure control. In conclusion, data on accidental or short-term exposure to hazardous chemicals are an important source of information for the further development of limit and assessment values, the validation of biomarkers and of targeted HBM programs for both routine monitoring and disaster management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Kaliumphosphat, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Sigma-Aldrich
4-Chloranilin, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Kaliumphosphat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Kaliumphosphat, tested according to Ph. Eur., anhydrous
Supelco
p-Chloranilin, Pharmaceutical Secondary Standard; Certified Reference Material