Merck
Sex differences in cardiovascular outcome during progression of aortic valve stenosis.
Heart (British Cardiac Society) (2014-10-11)
Dana Cramariuc, Barbara Patricia Rogge, Mai Tone Lønnebakken, Kurt Boman, Edda Bahlmann, Christa Gohlke-Bärwolf, John B Chambers, Terje R Pedersen, Eva Gerdts
ZUSAMMENFASSUNG
Women with severe aortic valve stenosis (AS) have better LV systolic function and more concentric LV geometry than their male counterparts. However, sex differences in cardiovascular (CV) outcome during progression of AS have not been reported from a longitudinal prospective study. Doppler echocardiography and CV events were recorded during a median of 4.0 years in 979 men and 632 women aged 28-86 (mean 67±10) years in the Simvastatin Ezetimibe in Aortic Stenosis (SEAS) study. LV systolic function was assessed by EF and midwall shortening (MWS). Study outcomes were AS-related events, ischaemic CV events and total mortality. The annular cumulative incidence of AS events, ischaemic CV events and death was 8.1%, 3.4% and 2.8% in women, and 8.9%, 4.4% and 2.4% in men, respectively. Women and men had similar AS progression rate whether measured by peak jet velocity, mean gradient or valve area. In multivariate analyses, female sex independently predicted less reduction in LV MWS and EF during follow-up (both p<0.05). In time-varying Cox analyses, women had a 40% lower rate of ischaemic CV events (95% CI 21% to 54%), in particular, more than 50% lower rate of stroke and coronary artery bypass grafting, and a 31% lower all-cause mortality (95% CI 1% to 51%), independent of active study treatment, age and hypertension, as well as time-varying valve area, low systolic function and abnormal LV geometry. AS event rate did not differ by sex. In the SEAS study, women and men had similar rates of AS progression and AS-related events. However, women had lower total mortality and ischaemic CV event rate than men independent of confounders. ClinicalTrials.gov identifier: NCT00092677.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Kreatinin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Simvastatin, analytical standard