Merck
Parenteral nutrition compromises neurodevelopment of preterm pigs.
The Journal of nutrition (2014-10-25)
Asim F Choudhri, Helen J Sable, Viktor V Chizhikov, Karyl K Buddington, Randal K Buddington
ZUSAMMENFASSUNG
Despite advances in nutritional support and intensive care, preterm infants are at higher risk of compromised neurodevelopment. This study evaluated the contribution of total parenteral nutrition (PN) to compromised neurodevelopment after preterm birth. Preterm pigs were provided PN or enteral nutrition (EN) for 10 d. Neurodevelopment was assessed by observations of motor activity and evaluation of sensory/motor reflexes, brain weight, MRI, and cerebellar histology. Despite similar gains in body weight, PN pigs had smaller brains (32 ± 0.4 vs. 35 ± 0.6 g; P = 0.0002) including the cerebellum, as well as reduced motor activity (P = 0.005), which corresponded to underdeveloped myelination (P = 0.004) measured by diffusion tensor imaging. PN resulted in lower serum triglycerides (17 ± 5.9 vs. 27 ± 3.1 mg/dL; P = 0.05), total cholesterol (31 ± 9.6 vs. 85 ± 8.1 mg/dL; P = 0.04), VLDL cholesterol (3.7 ± 1.2 vs. 5.7 ± 0.7 mg/dL; P = 0.04), and HDL cholesterol (16 ± 4.6 vs. 57 ± 7.3 mg/dL; P = 0.03) and nonsignificantly lower LDL cholesterol (10.7 ± 4.4 vs. 22.7 ± 2.9 mg/dL; P = 0.09). The compromised neurodevelopment caused by total PN is a novel finding, was independent of confounding variables (disease, inconsistent gestational ages, diverse genetics, extrauterine growth retardation, and inconsistent neonatal intensive care unit protocols), and highlights a need to improve current PN solutions. The preterm pig is a translational animal model for improving nutrition support to enhance neurodevelopment of preterm infants requiring PN.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Kreatinin, anhydrous, ≥98%
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Schweineniere, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Kälberdarmschleimhaut, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, alkalische Garnelen, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, alkalisch Rind, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein