Merck
Diabetic nephropathy is resistant to oral L-arginine or L-citrulline supplementation.
American journal of physiology. Renal physiology (2014-10-17)
Hanning You, Ting Gao, Timothy K Cooper, Sidney M Morris, Alaa S Awad
ZUSAMMENFASSUNG
Our recent publication showed that pharmacological blockade of arginases confers kidney protection in diabetic nephropathy via a nitric oxide (NO) synthase (NOS)3-dependent mechanism. Arginase competes with endothelial NOS (eNOS) for the common substrate L-arginine. Lack of L-arginine results in reduced NO production and eNOS uncoupling, which lead to endothelial dysfunction. Therefore, we hypothesized that L-arginine or L-citrulline supplementation would ameliorate diabetic nephropathy. DBA mice injected with multiple low doses of vehicle or streptozotocin (50 mg/kg ip for 5 days) were provided drinking water with or without L-arginine (1.5%, 6.05 g·kg(-1)·day(-1)) or L-citrulline (1.66%, 5.73 g·kg(-1)·day(-1)) for 9 wk. Nonsupplemented diabetic mice showed significant increases in albuminuria, blood urea nitrogen, glomerular histopathological changes, kidney macrophage recruitment, kidney TNF-α and fibronectin mRNA expression, kidney arginase activity, kidney arginase-2 protein expression, and urinary oxidative stress along with a significant reduction of nephrin and eNOS protein expression and kidney nitrite + nitrate compared with normal mice after 9 wk of diabetes. Surprisingly, L-arginine or L-citrulline supplementation in diabetic mice did not affect any of these parameters despite greatly increasing kidney and plasma arginine levels. These findings demonstrate that chronic L-arginine or L-citrulline supplementation does not prevent or reduce renal injury in a model of type 1 diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Perchlorsäure, ACS reagent, 70%
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
Perchlorsäure, 70%, 99.999% trace metals basis
Sigma-Aldrich
Perchlorsäure, ACS reagent, 60%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
SAFC
BIS-TRIS
Sigma-Aldrich
L-Citrullin, ≥98% (TLC)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Arginin, European Pharmacopoeia (EP) Reference Standard
Supelco
Perchlorsäure-Konzentrat, 0.01 M HClO4 in water (0.01N), eluent for IC
Supelco
4-tert.-Octylphenol-monoethoxylat -Lösung, 10 μg/mL in acetone, analytical standard