Merck
  • Phase 2 study of carfilzomib, thalidomide, and dexamethasone as induction/consolidation therapy for newly diagnosed multiple myeloma.
Phase 2 study of carfilzomib, thalidomide, and dexamethasone as induction/consolidation therapy for newly diagnosed multiple myeloma.
Blood (2014-11-16)
Pieter Sonneveld, Emilie Asselbergs, Sonja Zweegman, Bronno van der Holt, Marie Jose Kersten, Edo Vellenga, Marinus van Marwijk-Kooy, Annemiek Broyl, Okke de Weerdt, Sarah Lonergan, Antonio Palumbo, Henk Lokhorst
ZUSAMMENFASSUNG
This multicenter phase 2 study of the European Myeloma Network investigated the combination of carfilzomib, thalidomide, and dexamethasone (KTd) as induction/consolidation therapy for transplant-eligible patients with previously untreated multiple myeloma (N = 91). During KTd induction therapy, patients received 4 cycles of carfilzomib 20/27 mg/m(2) (n = 50), 20/36 mg/m(2) (n = 20), 20/45 mg/m(2) (n = 21), or 20/56 mg/m(2) (n = 20) on days 1, 2, 8, 9, 15, and 16 of a 28-day cycle; thalidomide 200 mg on days 1 to 28; and dexamethasone 20 mg on days 1, 2, 8, 9, 15, and 16. After autologous stem cell transplantation, patients proceeded to KTd consolidation therapy, where the target doses of carfilzomib were 27 mg/m(2), 36 mg/m(2), 45 mg/m(2), or 56 mg/m(2), respectively, and thalidomide 50 mg. Common grade 3/4 adverse events included respiratory (15%), gastrointestinal (12%), and skin disorders (10%); polyneuropathy was infrequent (1%). Complete response rates after induction and consolidation treatment were 25% and 63%, respectively; rates of very good partial response or better after induction and consolidation were 68% and 89%, respectively. At a median follow-up of 23 months, the 36-month progression-free survival rate was 72%. The KTd induction and consolidation regimens were active, safe, and well tolerated. This study was registered at http://www.trialregister.nl as #NTR2422.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethason – wasserlöslich, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Sigma-Aldrich
(±)-Thalidomid, ≥98%, powder
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard