Merck
  • Threonine 56 phosphorylation of Bcl-2 is required for LRRK2 G2019S-induced mitochondrial depolarization and autophagy.
Threonine 56 phosphorylation of Bcl-2 is required for LRRK2 G2019S-induced mitochondrial depolarization and autophagy.
Biochimica et biophysica acta (2014-12-03)
Yu-Chin Su, Xing Guo, Xin Qi
ZUSAMMENFASSUNG
The G2019S leucine-rich repeat kinase 2 (LRRK2) mutation is the most common cause of genetic Parkinson's disease (PD). However, the molecular mechanism underlying LRRK2 G2019S-induced cellular pathology is poorly understood. Here, we demonstrated that LRRK2 G2019S bound to and phosphorylated Bcl-2, a mitochondrial anti-apoptotic protein, at Threonine 56. Either stable expression of Bcl-2 or transient expression of a Bcl-2 phosphor mutant (Bcl-2(T56A)) abolished LRRK2 G2019S-induced mitochondrial depolarization and autophagy. Together, our findings reveal a previously unidentified target of LRRK2 G2019S, showing that Bcl-2 serves as a point of crosstalk between LRRK2 G2019S-mediated mitochondrial disorder and dysregulation of autophagy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
SAFC
L-Threonin
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonin, European Pharmacopoeia (EP) Reference Standard