Merck
Pax8 and Pax2 are specifically required at different steps of Xenopus pronephros development.
Developmental biology (2014-12-03)
Isabelle Buisson, Ronan Le Bouffant, Mélinée Futel, Jean-François Riou, Muriel Umbhauer
ZUSAMMENFASSUNG
The respective role of Pax2 and Pax8 in early kidney development in vertebrates is poorly understood. In this report, we have studied the roles of Pax8 and Pax2 in Xenopus pronephros development using a loss-of-function approach. Our results highlight a differential requirement of these two transcription factors for proper pronephros formation. Pax8 is necessary for the earliest steps of pronephric development and its depletion leads to a complete absence of pronephric tubule. Pax2 is required after the establishment of the tubule pronephric anlage, for the expression of several terminal differentiation markers of the pronephric tubule. Neither Pax2 nor Pax8 is essential to glomus development. We further show that Pax8 controls hnf1b, but not lhx1 and Osr2, expression in the kidney field as soon as the mid-neurula stage. Pax8 is also required for cell proliferation of pronephric precursors in the kidney field. It may exert its action through the wnt/beta-catenin pathway since activation of this pathway can rescue MoPax8 induced proliferation defect and Pax8 regulates expression of the wnt pathway components, dvl1 and sfrp3. Finally, we observed that loss of pronephros in Pax8 morphants correlates with an expanded vascular/blood gene expression domain indicating that Pax8 function is important to delimit the blood/endothelial genes expression domain in the anterior part of the dorso-lateral plate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
5-Brom-2′-deoxyuridin, ≥99% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
5-Brom-2′-deoxyuridin, BioUltra, ≥99%
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
USP
Dexamethason, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dexamethason, meets USP testing specifications
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason, British Pharmacopoeia (BP) Assay Standard
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard