Merck
Metal pollutants and cardiovascular disease: mechanisms and consequences of exposure.
American heart journal (2014-12-03)
Natalia V Solenkova, Jonathan D Newman, Jeffrey S Berger, George Thurston, Judith S Hochman, Gervasio A Lamas
ZUSAMMENFASSUNG
There is epidemiological evidence that metal contaminants may play a role in the development of atherosclerosis and its complications. Moreover, a recent clinical trial of a metal chelator had a surprisingly positive result in reducing cardiovascular events in a secondary prevention population, strengthening the link between metal exposure and cardiovascular disease (CVD). This is, therefore, an opportune moment to review evidence that exposure to metal pollutants, such as arsenic, lead, cadmium, and mercury, is a significant risk factor for CVD. We reviewed the English-speaking medical literature to assess and present the epidemiological evidence that 4 metals having no role in the human body (xenobiotic), mercury, lead, cadmium, and arsenic, have epidemiologic and mechanistic links to atherosclerosis and CVD. Moreover, we briefly review how the results of the Trial to Assess Chelation Therapy (TACT) strengthen the link between atherosclerosis and xenobiotic metal contamination in humans. There is strong evidence that xenobiotic metal contamination is linked to atherosclerotic disease and is a modifiable risk factor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quecksilber, ACS reagent, 99.9995% trace metals basis
Sigma-Aldrich
Blei, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Arsen, 99.999% trace metals basis
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Arsen, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Blei, shot, <2 mm, 99.9% trace metals basis
Blei, foil, 300x300mm, thickness 0.25mm, as rolled, 99.95%
Sigma-Aldrich
Blei, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Blei, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Blei, rod, 100mm, diameter 19.0mm, 99.95%
Blei, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Blei, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Blei, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Blei, foil, 50x50mm, thickness 1.0mm, as rolled, 99.5%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Blei, rod, 1000mm, diameter 3.2mm, 99.95%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Blei, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Blei, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Blei, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95%
Blei, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Blei, foil, 500x500mm, thickness 0.5mm, as rolled, 99.95%
Blei, rod, 100mm, diameter 6.35mm, 99.95%