Merck
  • Morphological alterations and acetylcholinesterase and monoamine oxidase inhibition in liver of zebrafish exposed to Aphanizomenon flos-aquae DC-1 aphantoxins.
Morphological alterations and acetylcholinesterase and monoamine oxidase inhibition in liver of zebrafish exposed to Aphanizomenon flos-aquae DC-1 aphantoxins.
Aquatic toxicology (Amsterdam, Netherlands) (2014-12-03)
De Lu Zhang, Jing Zhang, Chun Xiang Hu, Gao Hong Wang, Dun Hai Li, Yong Ding Liu
ZUSAMMENFASSUNG
Aphanizomenon flos-aquae is a cyanobacterium that produces neurotoxins or paralytic shellfish poisons (PSPs) called aphantoxins, which present threats to environmental safety and human health via eutrophication of water bodies worldwide. Although the molecular mechanisms of this neurotoxin have been studied, many questions remain unsolved, including those relating to in vivo hepatic neurotransmitter inactivation, physiological detoxification and histological and ultrastructural alterations. Aphantoxins extracted from the natural strain of A. flos-aquae DC-1 were analyzed by high-performance liquid chromatography. The main components were gonyautoxins 1 and 5 (GTX1, GTX5) and neosaxitoxin (neoSTX), which comprised 34.04%, 21.28%, and 12.77% respectively. Zebrafish (Danio rerio) were exposed intraperitoneally to 5.3 or 7.61 μg STX equivalents (eq)/kg (low and high doses, respectively) of A. flos-aquae DC-1 aphantoxins. Morphological alterations and changes in neurotransmitter conduction functions of acetylcholinesterase (AChE) and monoamine oxidase (MAO) in zebrafish liver were detected at different time points 1-24h post-exposure. Aphantoxin significantly enhanced hepatic alanine aminotransferase (ALT) and aspartate aminotransferase (AST) activities and histological and ultrastructural damage in zebrafish liver at 3-12 h post-exposure. Toxin exposure increased the reactive oxygen species content and reduced total antioxidative capacity in zebrafish liver, suggesting oxidative stress. AChE and MAO activities were significantly inhibited, suggesting neurotransmitter inactivation/conduction function abnormalities in zebrafish liver. All alterations were dose- and time-dependent. Overall, the results indicate that aphantoxins/PSPs induce oxidative stress through inhibition of AChE and MAO activities, leading to neurotoxicity in zebrafish liver. The above parameters may be useful as bioindicators for investigating aphantoxins/PSPs and cyanobacterial blooms in nature.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Osmiumtetroxid, ReagentPlus®, 99.8%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Osmiumtetroxid -Lösung, 4 wt. % in H2O
Sigma-Aldrich
Osmiumtetroxid -Lösung, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
USP
Eisessig, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Supelco
Essigsäure, analytical standard
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Osmiumtetroxid, ACS reagent, ≥98.0%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Supelco
5α-Androstan-17β-ol-3-on, VETRANAL®, analytical standard
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C