Merck
  • Fixed-dose rate gemcitabine alone or alternating with FOLFIRI.3 (irinotecan, leucovorin and fluorouracil) in the first-line treatment of patients with metastatic pancreatic adenocarcinoma: an AGEO randomised phase II study (FIRGEM).
Fixed-dose rate gemcitabine alone or alternating with FOLFIRI.3 (irinotecan, leucovorin and fluorouracil) in the first-line treatment of patients with metastatic pancreatic adenocarcinoma: an AGEO randomised phase II study (FIRGEM).
European journal of cancer (Oxford, England : 1990) (2014-12-03)
Isabelle Trouilloud, Anne-Claire Dupont-Gossard, David Malka, Pascal Artru, Mélanie Gauthier, Thierry Lecomte, Thomas Aparicio, Anne Thirot-Bidault, Céline Lobry, Amani Asnacios, Sophie Manet-Lacombe, Francine Fein, Olivier Dubreuil, Bruno Landi, Aziz Zaanan, Franck Bonnetain, Julien Taïeb
ZUSAMMENFASSUNG
Fluorouracil and irinotecan-based, and gemcitabine-based regimens, are the standard of care in the first-line treatment of patients with metastatic pancreatic cancer. New approaches are needed to improve survival and quality of life. Whether a sequential approach alternating irinotecan, fluorouracil and gemcitabine may be effective and tolerable in patients with metastatic pancreatic cancer is unknown. In this randomised, multicentre, open-label, phase 2 trial, patients with metastatic pancreatic adenocarcinoma, World Health Organisation (WHO) performance status 0-1, and bilirubin levels <1.5 upper limit of normal values (ULN) were randomised 1:1 to receive as first-line treatment either FOLFIRI.3 (irinotecan, leucovorin and fluorouracil) alternating with fixed-dose rate gemcitabine as 2-month periods (FIRGEM, arm A), or fixed-dose rate gemcitabine alone (arm B). Treatment was continued until disease progression or limiting toxicity. The primary end-point was the crude progression-free survival (PFS) rate at 6 months. The study is registered with EudraCT (N° 2006-005703-34). Between October 2007 and March 2011, 98 patients were enroled. The observed 6-month PFS rate was 43.5% (95% confidence interval (CI), [28.6-58.4%]) in arm A reaching the Fleming decision rules criteria to reject H0 and 26.1% (95% CI [12.9-39.3%]) in arm B. Objective response rates were 37% (23-51%) in arm A and 10% (1-19%) in arm B. Median PFS (5.0 versus 3.4 months, hazard ratio (HR)=0.59 [0.38-0.90]) and overall survival (11.0 versus 8.2 months, HR=0.71 [0.46-1.10]) were higher in arm A compared to arm B. The most frequent grade 3-4 toxicities were neutropenia (49%/24%; febrile neutropenia, 4%/0% in arms A/B), diarrhoea (arm A, 12% and arm B, 0%), and nausea/vomiting (8%/4%). No toxic deaths occurred. The FIRGEM strategy appears to be effective and feasible in patients with metastatic pancreatic cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Folinsäure Calciumsalz Hydrat, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
2′-Desoxycytidin, ≥99% (HPLC)
Supelco
Folinsäure Calciumsalz Hydrat, analytical standard
Sigma-Aldrich
Irinotecan -hydrochlorid, topoisomerase inhibitor
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Calciumfolinat, European Pharmacopoeia (EP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Fluorouracil, European Pharmacopoeia (EP) Reference Standard