Merck
  • Retrospective comparison of fludarabine in combination with intermediate-dose cytarabine versus high-dose cytarabine as consolidation therapies for acute myeloid leukemia.
Retrospective comparison of fludarabine in combination with intermediate-dose cytarabine versus high-dose cytarabine as consolidation therapies for acute myeloid leukemia.
Medicine (2014-12-17)
Wenjun Zhang, Yi Ding, Hao Wu, Yuhua Chen, Huina Lu, Chunying Chen, Jianfei Fu, Weiguang Wang, Aibin Liang, Shanhua Zou
ZUSAMMENFASSUNG
This retrospective study compared efficacy and safety of fludarabine combined with intermediate-dose cytarabine (FA regimen) versus high-dose cytarabine (HiDAC regimen) as consolidation therapy in acute myeloid leukemia (AML) patients who achieved complete remission. Disease-free survival (DFS) and overall survival (OS) based on age (≥ 60, <60 years) and cytogenetics were evaluated from data between January 2005 and March 2013. Total 82 patients (FA, n = 45; HiDAC, n = 37; 14-65 years) were evaluated. Five-year DFS was 32.0% and 36.2% for FA and HiDAC groups, respectively (P = 0.729), and 5-year OS was 39.5% and 47.8% (P = 0.568), respectively. Among older patients (≥ 60 years), 3-year DFS was 26.0% for FA group and 12.5% for HiDAC group (P = 0.032), and 3-year OS was 34.6% and 12.5%, respectively (P = 0.026). In FA group, hematological toxicities were significantly lower. FA regimen was as effective as HiDAC regimen in patients with good/intermediate cytogenetics and significantly improved DFS and OS in older patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Cytosin β-D-Arabinofuranosid -hydrochlorid, crystalline
Sigma-Aldrich
2-Fluoradenin-9-β-D-Arabinofuranosid, DNA synthesis and methylation inhibitor
Cytarabin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Adenin 9-β-D-Arabinofuranosid, ≥99%