Merck
Youngia erythrocarpa, a newly discovered cadmium hyperaccumulator plant.
Environmental monitoring and assessment (2014-12-17)
Lijin Lin, Bo Ning, Ming'an Liao, Yajun Ren, Zhihui Wang, Yingjie Liu, Ji Cheng, Li Luo
ZUSAMMENFASSUNG
The farmland weed Youngia erythrocarpa has been found to have the basic characteristics of a cadmium (Cd) hyperaccumulator. This study carried out preliminary and further Cd concentration gradient experiments and field experiment using Y. erythrocarpa to confirm this fact. The results showed that the biomass and resistance coefficient of Y. erythrocarpa decreased, but the root/shoot ratio and the Cd content in roots and shoots increased with the increase in soil Cd concentration. The Cd content in shoots of Y. erythrocarpa exceeded 100 mg/kg when the soil Cd concentration was 25 mg/kg in the two concentration gradient experiments, up to the maxima of 293.25 and 317.87 mg/kg at 100 mg/kg soil Cd. Both the bioconcentration factor of the shoots and the translocation factor exceeded 1 in all Cd treatments. In the field experiment, the total Cd extraction by shoots was 0.934-0.996 mg/m(2) at soil Cd levels of 2.04-2.89 mg/kg. Therefore, Y. erythrocarpa is a Cd hyperaccumulator that could be used to remediate Cd-contaminated farmland soil efficiently.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%
Cadmium, foil, 4mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, wire reel, 0.1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%