Merck
[How much coffee is healthy?].
Praxis (2014-12-11)
Franziska Etter
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
USP
Koffein, United States Pharmacopeia (USP) Reference Standard
Supelco
Schmelzpunktstandard 235-237°C, analytical standard
Supelco
Coffein Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Koffein -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Koffein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Koffein-Schmelzpunkt-Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Koffein, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Koffein, meets USP testing specifications, anhydrous
Supelco
Koffein -Lösung, analytical standard, 1.0 mg/mL in methanol
Supelco
Mettler Toledo Calibration Substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Koffein, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Koffein, BioXtra
Koffein für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Koffein, European Pharmacopoeia (EP) Reference Standard