Merck
Derivation and validation of homocysteine score in u.s. Men and women.
The Journal of nutrition (2014-12-21)
Seungyoun Jung, Youjin Je, Edward L Giovannucci, Bernard Rosner, Shuji Ogino, Eunyoung Cho
ZUSAMMENFASSUNG
One-carbon metabolism, which is crucial in DNA synthesis and genomic stability, is an interrelated network of biochemical reactions involved in several dietary and lifestyle factors. The development of the homocysteine score using these factors may be useful to reflect the status of one-carbon metabolism in large epidemiologic studies without biologic samples to measure homocysteine directly. The aim of this study was to develop an homocysteine score that reflects one-carbon metabolism better than individual dietary or lifestyle factors. We divided 2023 participants with measured plasma total homocysteine data in the Nurses' Health Study and the Health Professionals Follow-Up Study into training (n = 1619) and testing (n = 404) subsets. Using multivariable linear regression, we selected lifestyle determinants of plasma homocysteine in the training set and derived the homocysteine score weighted by the β coefficient for each predictor. The validation of the homocysteine score was assessed using the plasma homocysteine in the independent samples of the training set. In the training set, smoking, multivitamin use, and caffeine, alcohol, and dietary and supplemental folate intake were significant independent determinants of plasma homocysteine in multivariable linear regression (P ≤ 0.01) and were included in the derivation of the homocysteine score. The Pearson correlation of the homocysteine score with plasma homocysteine was 0.30 in the testing subset (P < 0.001). The homocysteine score was positively associated with the plasma homocysteine concentration in the testing subset and in an independent population of women; the mean difference of plasma homocysteine concentration between the extreme quintiles of homocysteine score ranged from 0.83 μmol/L to 1.52 μmol/L. Population misclassification either from the lowest quintile of plasma homocysteine into the highest quintile of the homocysteine score or from the highest quintile of plasma homocysteine into the lowest quintile of the homocysteine score was ≤12%. These data indicate that the homocysteine score may be used with relatively inexpensive and simple questionnaires to rank an individual's one-carbon metabolism status when homocysteine data are not available.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Koffein, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aktivkohle, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
Aktivkohle, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Folsäure, ≥97%
Sigma-Aldrich
Aktivkohle, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Folsäure, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Aktivkohle-Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Koffein, anhydrous, 99%, FCC, FG
Supelco
Folsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aktivkohle, untreated, granular, ≤5 mm
Sigma-Aldrich
Aktivkohle, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Aktivkohle, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Kohlenstoff, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
USP
Koffein, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Homocystein, ≥98.0% (NT)
Supelco
Schmelzpunktstandard 235-237°C, analytical standard
Supelco
Aktivkohle, powder
Sigma-Aldrich
Aktivkohle, acid-washed with hydrochloric acid
Sigma-Aldrich
Kohlenstoff, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Aktivkohle-Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Aktivkohle, meets USP testing specifications
Supelco
Aktivkohle, puriss. p.a., powder
Supelco
Coffein Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aktivkohle-Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Supelco
Koffein -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Folsäure, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aktivkohle, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Aktivkohle-Norit®, Norit® RB3, steam activated, rod
Supelco
Koffein, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland