Merck
Bedside methylene blue 'nephrostogram'.
Annals of the Royal College of Surgeons of England (2014-12-19)
S Pridgeon
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Methylenblau -Lösung, suitable for microscopy
Sigma-Aldrich
Methylenblau -Lösung, suitable for microbiology