Merck
  • A 44-year-old woman with metabolic acidosis, high anion gap, and delayed neurologic deterioration.
A 44-year-old woman with metabolic acidosis, high anion gap, and delayed neurologic deterioration.
Chest (2015-01-07)
Abhay Vakil, Hinesh Upadhyay, Khalid Sherani, Kelly Cervellione, Scott Trepeta, Mahendra C Patel
ZUSAMMENFASSUNG
A 44-year-old woman was brought to the ED from John F. Kennedy International Airport. The patient was returning with her son from a 3-month visit to Bangladesh. Her journey started with a 4-h flight from Dhaka, Bangladesh to Dubai, United Arab Emirates. She consumed 240 mL of whiskey during the flight. This was followed by a 14-h flight from Dubai to New York. According to the patient's son, she did not consume any alcohol during the second flight. The patient was in her usual state of health with normal mentation throughout her journey. Upon landing, she started complaining of shortness of breath. After disembarking, she was witnessed to have seizure-like activity with involuntary passage of urine, following which she collapsed. The patient was intubated by emergency medical services in the field.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.