Merck
  • Online mindfulness-based intervention for late-stage bipolar disorder: pilot evidence for feasibility and effectiveness.
Online mindfulness-based intervention for late-stage bipolar disorder: pilot evidence for feasibility and effectiveness.
Journal of affective disorders (2015-03-22)
G Murray, N D Leitan, M Berk, N Thomas, E Michalak, L Berk, S L Johnson, S Jones, T Perich, N B Allen, Michael Kyrios
ZUSAMMENFASSUNG
People in the late stage of bipolar disorder (BD) experience elevated relapse rates and poorer quality of life (QoL) compared with those in the early stages. Existing psychological interventions also appear less effective in this group. To address this need, we developed a new online mindfulness-based intervention targeting quality of life (QoL) in late stage BD. Here, we report on an open pilot trial of ORBIT (online, recovery-focused, bipolar individual therapy). Inclusion criteria were: self-reported primary diagnosis of BD, six or more episodes of BD, under the care of a medical practitioner, access to the internet, proficient in English, 18-65 years of age. Primary outcome was change (baseline - post-treatment) on the Brief QoL.BD (Michalak and Murray, 2010). Secondary outcomes were depression, anxiety, and stress measured on the DASS scales (Lovibond and Lovibond, 1993). Twenty-six people consented to participate (Age M=46.6 years, SD=12.9, and 75% female). Ten participants were lost to follow-up (38.5% attrition). Statistically significant improvement in QoL was found for the completers, t(15)=2.88, 95% CI:.89-5.98, p=.011, (Cohen׳s dz=.72, partial η(2)=.36), and the intent-to-treat sample t(25)=2.65, 95% CI:.47-3.76, (Cohen׳s dz=.52; partial η(2)=.22). A non-significant trend towards improvement was found on the DASS anxiety scale (p=.06) in both completer and intent-to-treat samples, but change on depression and stress did not approach significance. This was an open trial with no comparison group, so measured improvements may not be due to specific elements of the intervention. Structured diagnostic assessments were not conducted, and interpretation of effectiveness was limited by substantial attrition. Online delivery of mindfulness-based psychological therapy for late stage BD appears feasible and effective, and ORBIT warrants full development. Modifications suggested by the pilot study include increasing the 3 weeks duration of the intervention, adding cautions about the impact of extended meditations, and addition of coaching support/monitoring to optimise engagement.