Merck
  • BDNF-TrkB signaling through Erk1/2 MAPK phosphorylation mediates the enhancement of fear memory induced by glucocorticoids.
BDNF-TrkB signaling through Erk1/2 MAPK phosphorylation mediates the enhancement of fear memory induced by glucocorticoids.
Molecular Psychiatry (2014)
Revest JM, Le Roux A, Roullot-Lacarriere V, et al.