Merck
  • Lipopolysaccharide-induced microRNA-146a targets CARD10 and regulates angiogenesis in human umbilical vein endothelial cells.
Lipopolysaccharide-induced microRNA-146a targets CARD10 and regulates angiogenesis in human umbilical vein endothelial cells.
Toxicological sciences : an official journal of the Society of Toxicology (2014-05-28)
Cheng-Shyuan Rau, Johnson Chia-Shen Yang, Yi-Chun Chen, Chia-Jung Wu, Tsu-Hsiang Lu, Siou-Ling Tzeng, Yi-Chan Wu, Ching-Hua Hsieh
ZUSAMMENFASSUNG
This aim of this study was to explore the role of miRNA-146a (miR-146a) and its target genes in endothelial cells. We demonstrated that lipopolysaccharide (LPS) induced the upregulation of miR-146a in human umbilical vein endothelial cells (HUVECs), and that the induction was blocked by silencing toll-like receptors, the adaptor molecule MyD88, and the nonspecific NF-κB inhibitor BAY 11-7082. In addition, knockdown of miR-146a by transfection of the locked nucleic acid antimiR-146a significantly inhibited LPS-induced cell migration and tube formation. A combined analysis of bioinformatics miRanda algorithms and a whole genome expression microarray of immunoprecipitated Ago2 ribonucleoprotein complexes identified 14 potential target genes. Subsequent transfection with the miR-146a precursor pre-miR-146a into HUVECs validated that CARD10 was the target gene of the miR-146a, both at the mRNA and protein levels. Silencing CARD10 inhibited p65 nuclear translocation in the cells receiving LPS stimulation and increased angiogenesis. Therefore, miR-146a may play a role in regulating the angiogenesis in HUVECs by downregulating CARD10, which acts in a negative feedback regulation loop to inhibit the activation of NF-κB that normally impairs angiogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumbicarbonat, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
USP
Natriumbicarbonat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumbicarbonat, −40-+140 mesh, ≥95%
Sigma-Aldrich
Natriumbicarbonat, tested according to Ph. Eur.
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O26:B6, purified by trichloroacetic acid extraction
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting human CARD10