Merck
  • An ITAM-Syk-CARD9 signalling axis triggers contact hypersensitivity by stimulating IL-1 production in dendritic cells.
An ITAM-Syk-CARD9 signalling axis triggers contact hypersensitivity by stimulating IL-1 production in dendritic cells.
Nature Communications (2014)
Yasukawa S, Miyazaki Y, Yoshii C, et al.