Merck
  • Global analysis of p53-regulated transcription identifies its direct targets and unexpected regulatory mechanisms.
Global analysis of p53-regulated transcription identifies its direct targets and unexpected regulatory mechanisms.
eLife (2014)
Allen MA, Andrysik Z, Dengler VL, et al.