Merck
Cerebral vascular leak in a mouse model of amyloid neuropathology.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism (2014-07-24)
Eric A Tanifum, Zbigniew A Starosolski, Stephanie W Fowler, Joanna L Jankowsky, Ananth V Annapragada
ZUSAMMENFASSUNG
In Alzheimer's disease (AD), there is increasing evidence of blood-brain barrier (BBB) compromise, usually observed as 'microbleeds' correlated with amyloid plaque deposition and apoE-ɛ4 status, raising the possibility of nanotherapeutic delivery. Molecular probes have been used to study neurovascular leak, but this approach does not adequately estimate vascular permeability of nanoparticles. We therefore characterized cerebrovascular leaks in live APP+ transgenic animals using a long circulating ∼100 nm nanoparticle computed tomography (CT) contrast agent probe. Active leaks fell into four categories: (1) around the dorsomedial cerebellar artery (DMCA), (2) around other major vessels, (3) nodular leaks in the cerebral cortex, and (4) diffuse leaks. Cortical leaks were uniformly more frequent in the transgenic animals than in age-matched controls. Leaks around vessels other than the DMCA were more frequent in older transgenics compared with younger ones. All other leaks were equally prevalent across genotypes independent of age. Ten days after injection, 4 to 5 μg of the dose was estimated to be present in the brain, roughly a half of which was in locations other than the leaky choroid plexus, and associated with amyloid deposition in older animals. These results suggest that amyloid deposition and age increase delivery of nanoparticle-borne reagents to the brain, in therapeutically relevant amounts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Gelatine aus Schweinehaut, gel strength 300, Type A
Sigma-Aldrich
Oxalsäure, 98%
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Sigma-Aldrich
Oxalsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumdisulfid, ≥98%
Sigma-Aldrich
Thioflavin S, practical grade
Sigma-Aldrich
Oxalsäure, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalsäure, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Supelco
o-Xylen, analytical standard
Supelco
o-Xylen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Oxalsäure-Konzentrat, 0.1 M (COOH)2 (0.2N), eluent concentrate for IC