Merck
  • The cAMP-responsive element binding protein (CREB) transcription factor regulates furin expression during human trophoblast syncytialization.
The cAMP-responsive element binding protein (CREB) transcription factor regulates furin expression during human trophoblast syncytialization.
Placenta (2014)
Zhou Z, Wang R, Yang X, et al.