Merck
  • Exploring the potential of xanthene derivatives as trypanothione reductase inhibitors and chloroquine potentiating agents.
Exploring the potential of xanthene derivatives as trypanothione reductase inhibitors and chloroquine potentiating agents.
Tetrahedron (2003)
Chibale K, et al