Merck
  • Houben-Weyl Methods of Molecular Transformations. Heteroatom Analogues of Aldehydes and Ketones.
Houben-Weyl Methods of Molecular Transformations. Heteroatom Analogues of Aldehydes and Ketones.
Science of Synthesis Knowledge Updates (2014)
Aggarwal VK