Merck
AMPK is a direct adenylate charge-regulated protein kinase.
Science (New York, N.Y.) (2011-06-18)
Jonathan S Oakhill, Rohan Steel, Zhi-Ping Chen, John W Scott, Naomi Ling, Shanna Tam, Bruce E Kemp
ZUSAMMENFASSUNG
The adenosine monophosphate (AMP)-activated protein kinase (AMPK) regulates whole-body and cellular energy balance in response to energy demand and supply. AMPK is an αβγ heterotrimer activated by decreasing concentrations of adenosine triphosphate (ATP) and increasing AMP concentrations. AMPK activation depends on phosphorylation of the α catalytic subunit on threonine-172 (Thr(172)) by kinases LKB1 or CaMKKβ, and this is promoted by AMP binding to the γ subunit. AMP sustains activity by inhibiting dephosphorylation of α-Thr(172), whereas ATP promotes dephosphorylation. Adenosine diphosphate (ADP), like AMP, bound to γ sites 1 and 3 and stimulated α-Thr(172) phosphorylation. However, in contrast to AMP, ADP did not directly activate phosphorylated AMPK. In this way, both ADP/ATP and AMP/ATP ratios contribute to AMPK regulation.