Merck
  • Reaction of a vesicular functionalized surfactant with 2-chloroethyl phenyl sulfide, a mustard simulant.
Reaction of a vesicular functionalized surfactant with 2-chloroethyl phenyl sulfide, a mustard simulant.
Langmuir (1999)
Jaeger DA, et al.