Merck
  • The synthesis and structural characterization of [Et4N]2 [Ti(SPh)6] and trimeric [Ti3O(SPh)3Cl4 (CH3 CN)5]?CH3 CN?(C2 H5)2O complexes.
The synthesis and structural characterization of [Et<SUB>4</SUB>N]<SUB>2</SUB> [Ti(SPh)<SUB>6</SUB>] and trimeric [Ti<SUB>3</SUB>O(SPh)<SUB>3</SUB>Cl<SUB>4</SUB> (CH<SUB>3</SUB> CN)<SUB>5</SUB>]?CH<SUB>3</SUB> CN?(C<SUB>2</SUB> H<SUB>5</SUB>)<SUB>2</SUB>O complexes.
Polyhedron (2000)
Kim JT, et al.