Merck
  • Preparation of pyrrole and pyrrolidine derivatives of pyrimidine. 1-(2-pyrimidinyl) pyrrole-an inhibitor of X. phaseoli and X. malvacearum.
Preparation of pyrrole and pyrrolidine derivatives of pyrimidine. 1-(2-pyrimidinyl) pyrrole-an inhibitor of <I>X. phaseoli and X. malvacearum</I>.
Journal of Heterocyclic Chemistry (2004)
Becker I.