Merck
  • Ammonium chloride-catalyzed one-pot synthesis of 3,4-dihydropyrimidin-2-(1H)-ones under solvent-free conditions.
Ammonium chloride-catalyzed one-pot synthesis of 3,4-dihydropyrimidin-2-(1<I>H</I>)-ones under solvent-free conditions.
Tetrahedron Letters (2003)
Shaabani A, et al.