- Tackling writer's block. Suggestions for self-modification.
Tackling writer's block. Suggestions for self-modification.
The Journal of nursing administration (1985-05-01)
H Rosenberg, M I Lah
PMID3845971
ZUSAMMENFASSUNG
Writing is an important and pervasive part of every administrator's duties, and problems with writer's block and procrastination can impede one's productivity. This article describes behavioral assessment and intervention methods (e.g., task analysis, reinforcement analysis, time-structuring, charting, and deposit contracting) that an individual can use to modify his or her writing habits.
MATERIALIEN