Merck
  • Identification of PDZK4, a novel human gene with PDZ domains, that is upregulated in synovial sarcomas.
Identification of PDZK4, a novel human gene with PDZ domains, that is upregulated in synovial sarcomas.
Oncogene (2004-04-13)
Satoshi Nagayama, Megumi Iiizumi, Toyomasa Katagiri, Junya Toguchida, Yusuke Nakamura
ZUSAMMENFASSUNG
In an earlier study designed to investigate molecular mechanisms of carcinogenesis in synovial sarcomas (SSs), we applied a cDNA microarray to detect human genes with significantly increased expression in SS cells. Among the genes selected in this way, we identified a novel transcript, subsequently designated PDZK4 (PDZ domain-containing 4), that was specifically upregulated in all of the 13 SS cases we examined. On Northern blots of normal human tissues, the PDZK4 transcript was expressed only in fetal brain. Immunocytochemical staining of transfected COS7 cells showed that the PDZK4 gene product localized mainly under the plasma membrane. Treatment of human SS cells with small interfering RNA (siRNA) inhibited the expression of PDZK4 and resulted in the suppression of tumor-cell growth. Induction of exogenous PDZK4 expression promoted growth of T98G and COS7 cells in which no endogenous expression of PDZK4 was observed. Taken together, these findings strongly suggest that inappropriate expression of PDZK4 might play an important role in the proliferation of SS cells and that the gene might be a suitable molecular target for designing of novel drugs to treat SS patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid