Merck
  • Zirconium(IV) Chloride - Catalysed Reaction of Indoles: An Expeditious Synthesis of Bis(indolyl)methanes
Zirconium(IV) Chloride - Catalysed Reaction of Indoles: An Expeditious Synthesis of Bis(indolyl)methanes
Bull. Korean Chem. Soc. (2005)
Nagawade RR, et al.