Merck
  • A simple and effective method to encapsulate tobacco mesophyll protoplasts to maintain cell viability
A simple and effective method to encapsulate tobacco mesophyll protoplasts to maintain cell viability
Methods (2015)
Lei R, et al.