Merck
  • Human Zwint-1 specifies localization of Zeste White 10 to kinetochores and is essential for mitotic checkpoint signaling.
Human Zwint-1 specifies localization of Zeste White 10 to kinetochores and is essential for mitotic checkpoint signaling.
The Journal of biological chemistry (2004-10-16)
Hongmei Wang, Xiaoyu Hu, Xia Ding, Zhen Dou, Zhihong Yang, Andrew W Shaw, Maikun Teng, Don W Cleveland, Michael L Goldberg, Liwen Niu, Xuebiao Yao
ZUSAMMENFASSUNG
Chromosome segregation in mitosis is orchestrated by dynamic interaction between spindle microtubules and the kinetochore, a multiprotein complex assembled onto centromeric DNA of the chromosome. Here we show that Zwint-1 is required and is sufficient for kinetochore localization of Zeste White 10 (ZW10) in HeLa cells. Zwint-1 specifies the kinetochore association of ZW10 by interacting with its N-terminal domain. Suppression of synthesis of Zwint-1 by small interfering RNA abolishes the localization of ZW10 to the kinetochore, demonstrating the requirement of Zwint-1 for ZW10 kinetochore localization. In addition, depletion of Zwint-1 affects no mitotic arrest but causes aberrant premature chromosome segregation. These Zwint-1-suppressed cells display chromosome bridge phenotype with sister chromatids inter-connected. Moreover, Zwint-1 is required for stable association of CENP-F and dynamitin but not BUB1 with the kinetochore. Finally, our studies show that Zwint-1 is a new component of the mitotic check-point, as cells lacking Zwint-1 fail to arrest in mitosis when exposed to microtubule inhibitors, yielding interphase cells with multinuclei. As ZW10 and Zwint-1 are absent from yeast, we reasoned that metazoans evolved an elaborate spindle checkpoint machinery to ensure faithful chromosome segregation in mitosis.